Learn More
Poulten Selfe™ Glass Routine Viscometer
Product Code. 13181073
Change view
Click to view available options
Viscosity Range:
20 to 100 cSt
Unit Size:
Each
Product Code. Viscosity Range Unit Size
13181073 20 to 100 cSt Each
1 options
This item is not returnable. View return policy
Product Code. 13181073 Supplier Poulten Selfe™ Supplier No. 1634/06
  • ONLINE EXCLUSIVE PRICE
    178.00€
    172.97€ / paquete de 1
    Ahorro 5.03€
    2% Descuento
 Restricted Item
Fecha estimada de envoi: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for measuring transparent liquids
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids, nominal overall length 250mm. Approximate sample size 7mL. Calibration data at 40°C with the constant, C, stated at 40°C and 100°C. With NAMAS certification of calibration.
Specifications
Product Type Routine Viscometer
Viscosity Range 20 to 100 cSt
Calibrated Yes
Material Glass
Size 200