Learn More
Macherey-Nagel™ Glass Plate Adamant
Product Code. 12814431 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
5 x 10 cm
10 x 10 cm
20 x 20 cm
Quantity:
200 Pk.
25 Pk.
50 Pk.
Unit Size:
Pack of 200
Pack of 25
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity Unit Size
12814431 5 x 10 cm 200 Pk. Pack of 200
10181802 10 x 10 cm 25 Pk. Pack of 25
10264522 20 x 20 cm 25 Pk. Pack of 25
10356022 5 x 10 cm 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12814431 Supplier Macherey-Nagel™ Supplier No. 821040.200
  • ONLINE EXCLUSIVE PRICE
    338.00€
    292.17€ / paquete de 200
    Ahorro 45.83€
    13% Descuento
Fecha estimada de envoi: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 200 Pk.
Type Glass Plate
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Dimensions (L x W) 5 x 10 cm