Learn More
Brady™ Glass-Filled Nylon Miniature Circuit Breaker Lockouts
Product Code. 15623517 Shop All Brady Products
Change view
Click to view available options
Color:
Red
Red/Black
Unit Size:
1 pack
Each
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15623517 Red Pack of 6
11731356 Red 1 pack
11771945 Red/Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15623517 Supplier Brady™ Supplier No. 090854
  • ONLINE EXCLUSIVE PRICE
    134.00€
    114.67€ / paquete de 6
    Ahorro 19.33€
    14% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
A safe and effective method for locking out European Circuit Breakers.
  • Requires no tools to install
  • Lockout easily installs with the aid of a push button
  • Tie-bar style lockout has a thumb wheel for quick application
Specifications
Product Type Miniature Circuit Breaker Lockouts
Alternate Part Number ELEC. CIRCUIT BREAKER L/O EURO TB PK6
Color Red
Material Glass-filled Nylon
Description Miniature Circuit Breaker Lockouts - Tie-Bar
Diameter (Metric) Shackle 7 mm
Dimensions (D x W x H) 24.13 x 31.24 x 54.9 mm
Material (Shackle) Glass-filled Nylon
Quantity 6 Pk.
Width (Metric) 31.24 mm