Learn More
Labconco™ Glass Chamber Lids
Product Code. 10454092 Shop All Labconco Corporation Products
Change view
Click to view available options
Diameter (Metric):
24 cm
Unit Size:
Each
Product Code. Diameter (Metric) Unit Size
10454092 24 cm Each
1 options
This item is not returnable. View return policy
Product Code. 10454092 Supplier Labconco™ Supplier No. 7439900
  • ONLINE EXCLUSIVE PRICE
    815.00€
    673.57€ / paquete de 1
    Ahorro 141.43€
    17% Descuento
 Restricted Item
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For collector and drying chambers.
  • Recommended for applications in which solvents that damage acrylic are used
Specifications
Type Chamber Lid
Material Tempered Glass
Size Small
For Use With (Application) Provides additional protection from solvents and corrosives that attach acrylic, such as acetonitrile
For Use With (Equipment) Freezone™ 4.5l freeze-dry system collectors/drying chambers or 12-port drying chamber
Includes Rubber gasket
Diameter (English) 9.5 in.
Diameter (Metric) 24 cm
Weight (English) 2 lb.
Weight (Metric) 0.9 kg