missing translation for 'onlineSavingsMsg'
Learn More

GIPR Antibody (Gipg013), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-81067JF646

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216309

  • 595.02€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

GIPR Monoclonal specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

GIPR
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
GIPR
Human GIPR.
0.1 mL
GPCR, Neuroscience
2696
Human, Mouse, Rat, Canine
Purified
Immunofluorescence
Gipg013
Immunocytochemistry/Immunofluorescence
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Mouse
Protein A purified
RUO
Primary
The original has a Ki value of 7 nM and a Kd value of 6.8 nM for human GIPR.
Store at 4°C in the dark.
IgG2a λ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales