Learn More
Gilac™ Gilactiv™ Tumblers, 22 Cycle, Blue
Product Code. 17978961
Change view
Click to view available options
Capacity (Metric):
0.22 L
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
17978961 0.22 L Each
1 options
This item is not returnable. View return policy
Product Code. 17978961 Supplier Gilac™ Supplier No. G627290
  • ONLINE EXCLUSIVE PRICE
    15.21€
    10.05€ / paquete de 1
    Ahorro 5.16€
    33% Descuento
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Gilactiv™ 22 Cycle Tumblers are reusable plastic tumblers treated with the exclusive Sanitized™ technology, which protects the tumbler's surface against viruses and bacteria.
  • Gilactiv™ reusable plastic tumbler treated with the exclusive Sanitized™ technology, which protects the tumbler's surface against viruses and bacteria.
  • Proven antibacterial and antiviral effectiveness on the tumbler's surface (empty or full) for a broad spectrum of germs, including HCoV-229E human coronavirus.
  • This article contains a biocidal product with the active substance: Silver Phosphate Glass.
  • Tests conducted by an EU-approved French independent laboratory showed that 98.6% of the virus was wiped out after a two-hour period.
  • Choose Gilactiv™ enhanced protection.
    • Test results available on request.
  • Outstanding durability even in the intensive use cycle.
Specifications
Material Plastic
Capacity (Metric) 0.22 L
Capacity (English) 7.4 oz.
Quantity 10 Pk.
Product Type Tumbler Cup