Learn More
Gibco™ Human NT-4 Recombinant Protein
Human NT-4 Recombinant Protein
Marca: Gibco™ PHC7024
Detalles adicionales : Peso : 0.50000kg
Descripción
Human NT-4 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
NT-4 is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Especificaciones
Cell Culture | |
27.2 kDa | |
5 ug | |
260 aa (homodimer) | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Stem Cell Research, Differentiation, Signal Transduction | |
98 % |
4909 | |
Sequential Chromatography | |
NT-4 | |
Human | |
Carrier-Free | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
Neurotrophic Factors | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.