missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse CXCL10 (IP-10) Recombinant Protein
Recombinant IP10 is a bioactive protein intended for use in cell culture applications
en la oferta
Especificaciones
Método de purificación | Sequential Chromatography |
---|---|
Alias de gen | IP-10 |
ID de gen (Entrez) | 15945 |
Descripción
IP10 is a chemokine of the CXC subfamily and ligand for the receptor CXCR3. Binding of this protein to CXCR3 results in pleiotropic effects, including stimulation of monocytes, natural killer and T-cell migration, and modulation of adhesion molecule expression.Approved for shipment on Wet or Dry Ice.
Especificaciones
Sequential Chromatography | |
IP-10 | |
15945 |
For Research Use Only. Not for use in diagnostic procedures.