Learn More
Gibco™ IL13 Recombinant Mouse Protein
Recombinant Mouse Interleukin 13 (IL13) is a bioactive protein intended for use in cell culture applications
Marca: Gibco™ PMC0131
Detalles adicionales : Peso : 0.02000kg
Descripción
IL13 down-regulates macrophage produced cytokines such as TNF-α, IL-1, IL-6, IL-8, and IL-12 in response to LPS challenge or IFN-γ. IL13 induces monocyte differentiation and also induces B cell differentiation and proliferation.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human TF-1 cells
Shipping Condition: Wet Ice.
Especificación
Cell Culture; Mammalian Cell Culture | |
16163 | |
Sequential Chromatography | |
100μg | |
Store at 2° to 8°C. | |
Recombinant/Ligand | |
<0.1ng/μg | |
Cell Culture | |
E. coli |
Lyophilized | |
12kDa | |
98% | |
Mouse | |
Carrier Protein | |
Interleukins | |
IL-13 | |
Cytokines & Receptors |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.