missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ GM-CSF Recombinant Rat Protein

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

Marca:  Gibco™ PRC2014

Detalles adicionales : Peso : 0.05250kg

 Ver más versiones de este producto

Código de producto. 10425723

  • 134.00€ / cada uno


Fecha de envío estimada
del proveedor 11-02-2019
Añadir a la cesta

Descripción

Descripción

Recombinant rat GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Approved for shipment on Wet or Dry Ice.

Especificación

Especificación

Cell Culture; Mammalian Cell Culture
116630
Exchange Chromatography
2μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
14.6kDa
95%
Rat
Carrier Protein
GM-CSF, G-CSF, & M-CSF
GM-CSF
Cytokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.