Learn More
Gibco™ Human FGF-18 Recombinant Protein
Human FGF-18 Recombinant Protein
Marca: Gibco™ PHG0234
Detalles adicionales : Peso : 0.50000kg
Descripción
Human FGF-18 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
FGF18 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. Studies suggest that this protein is a pleiotropic growth factor that stimulates proliferation in a number of tissues, most notably the liver and small intestine.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Especificaciones
Cell Culture | |
21.2 kDa | |
25 ug | |
Human | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis | |
95 % |
8817 | |
Sequential Chromatography | |
FGF-18 | |
Store in freezer (-5 to -30°C). | |
25 μg | |
E. coli | |
Recombinant, Ligand | |
FGFs (Fibroblast Growth Factors) | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.