Learn More
Gibco™ Human CXCL2 (GRO beta) Recombinant Protein
Human CXCL2 (GRO beta) Recombinant Protein
Marca: Gibco™ PHC1074
Detalles adicionales : Peso : 0.50000kg
Descripción
Human CXCL2 (GRO beta) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl
CXCL2 is a chemokine that induces the infiltration of neutrophils.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Especificaciones
Cell Culture | |
Sequential Chromatography | |
CXCL2 (GROβ) | |
Human | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Chemokines & Receptors | |
Lyophilized | |
Inflammation, Oncology | |
98 % |
7.9 kDa | |
10 ug | |
73 aa | |
Store in freezer (-5 to -30°C). | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
GRO⁄KC⁄CINC Chemokines | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.