Learn More
Gibco™ TNF Recombinant Human Protein
TNF; Recombinant Human Protein
Marca:  Gibco™ CTP3013
Detalles adicionales : Peso : 0.99000kg
Código de producto. 10630301
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Requires : Recombinant TNF is a bioactive protein intended for use in cell culture applications. TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.
Descripción
Descripción
  • TNF causes cytolysis and cytostasis of many tumor cell lines
  • With wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis
  • ED50 range = 0.02 to 0.05ng/mL (Specific Activity: 2 x 107 to 5 x 107 units/mg), determined by dose dependent cytotoxic effect on L929 cells in presence of actinomycin D
Cell Culture, Dendritic Cell Culture, Hematopoietic Stem Cell Culture, Mammalian Cell Culture, Mesenchymal Stem Cell Culture, Primary Cell Culture, Stem Cell Culture, Stem Cell Research
Información de órdenes
Shipping Condition: Wet ice
Especificaciones
Especificaciones
1 mg
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only. Not for use in diagnostic procedures.