missing translation for 'onlineSavingsMsg'
Learn More

GFAP (Astrocyte & Neural Stem Cell Marker) Mouse, Clone: GFAP/2076, enQuireBio™

Mouse Monoclonal Antibody

Marca:  EnquireBio 2670-MSM6-P0

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15910088

  • 214.00€ / 20 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

GFAP (Astrocyte & Neural Stem Cell Marker)
Cytoplasmic
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human GFAP protein fragment (apprx. aa 101-200)
∽50kDa
Protein A/G purified from Bioreactor concentrate.
RUO
Primary
2670
Western Blot, Immunohistochemistry
GFAP/2076
Unconjugated
Mouse
IgG1
Brain or Astrocytoma
20 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos