missing translation for 'onlineSavingsMsg'
Learn More

Geranylacetone, (E)+(Z), 97%, (Z)-isomer (nerylacetone) ca 45%

Product Code. 11419466
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419466 100 g 100g
11409466 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419466 Supplier Thermo Scientific Alfa Aesar Supplier No. A19184.22
 Restricted Item
Fecha estimada de envoi: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 689-67-8 | C13H22O | 194.318 g/mol

It is used as a flavoring agent and adjuvant. It is also employed as a substance, extract, or preparation for diffusing or imparting an agreeable or attractive smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a flavoring agent and adjuvant. It is also employed as a substance, extract, or preparation for diffusing or imparting an agreeable or attractive smell.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refregirated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 689-67-8
Molecular Formula C13H22O
Molecular Weight (g/mol) 194.318
MDL Number MFCD00008910
InChI Key HNZUNIKWNYHEJJ-FMIVXFBMSA-N
Synonym geranylacetone, geranyl acetone, e-6,10-dimethylundeca-5,9-dien-2-one, trans-geranylacetone, 5e-6,10-dimethylundeca-5,9-dien-2-one, geranylacetone, trans, dihydropseudoionone, 6,10-dimethylundeca-5,9-dien-2-one, e-6,10-dimethyl-5,9-undecadien-2-one, unii-9b7ry79u9z
PubChem CID 1549778
ChEBI CHEBI:67206
IUPAC Name (5E)-6,10-dimethylundeca-5,9-dien-2-one
SMILES CC(=CCCC(=CCCC(=O)C)C)C

Specifications

Density 0.87
Boiling Point 250°C
Flash Point 110°C (230°F)
Refractive Index 1.467
Quantity 100 g
Beilstein 1722277
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 194.32
Percent Purity 97%
Assay (Z)-isomer (nerylacetone) ≈45%
Chemical Name or Material Geranylacetone, (E)+(Z)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.