Learn More
Thermo Scientific™ Fiber Optic Probes for GENESYS™
Product Code. 15896095 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
GENESYS G5
GENESYS G5 and Evolution Pro
GENESYS G5, Evolution One Plus, and Evolution Pro spectrophtometers
Type:
Fiber Optic Coupler
Fiber Optic Dip Probe System
Fiber Optic Dip Probe, 10 mm
Tip for Dip Probe, 10 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Unit Size
15896095 GENESYS G5 Fiber Optic Dip Probe System Each
15841148 GENESYS G5 and Evolution Pro Fiber Optic Dip Probe, 10 mm Each
15806105 GENESYS G5 Fiber Optic Coupler Each
15861148 GENESYS G5, Evolution One Plus, and Evolution Pro spectrophtometers Tip for Dip Probe, 10 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15896095 Supplier Thermo Scientific™ Supplier No. 840305700
  • ONLINE EXCLUSIVE PRICE
    5175.00€
    4819.50€ / paquete de 1
    Ahorro 355.50€
    6% Descuento
Fecha estimada de envoi: 13-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurement requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Preserve sample integrity while minimizing sample carryover.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 6 mm
Type Fiber Optic Dip Probe System
For Use With (Equipment) GENESYS G5