missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEMIN6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12307-2-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
GEMIN6 is part of a large macromolecular complex, localized to both the cytoplasm and the nucleus, that plays a role in the cytoplasmic assembly of small nuclear ribonucleoproteins (snRNPs). Other members of this complex include SMN (MIM 600354), GEMIN2 (SIP1; MIM 602595), GEMIN3 (DDX20; MIM 606168), GEMIN4 (MIM 606969), and GEMIN5 (MIM 607005).Especificaciones
GEMIN6 | |
Polyclonal | |
Unconjugated | |
GEMIN6 | |
Gem associated protein 6, Gemin 6, GEMIN6, SIP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362688, 67242, 79833 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WXD5, Q9CX53 | |
Gemin6 | |
GEMIN6 Fusion Protein Ag2954 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |