missing translation for 'onlineSavingsMsg'
Learn More

Gelsolin Mouse, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

533.68€

Especificaciones

Antígeno Gelsolin
Clon 2
Concentración 250μg/mL
Aplicaciones Western Blot
Clasificación Monoclonal
Ver más especificaciones

Productos 1
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15835449
Ver documentos Detalles de la promoción
BD Biosciences
610413
150 μg Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A Ver productos alternativos
Ahorre hasta 
»
Descripción

Descripción

Gelsolin was identified as a result of its ability to sever actin filaments in a Ca2+-dependent manner. The gene for gelsolin encodes an 83 kDa protein that migrates as a 93 kDa polypeptide in SDS-gels. The N-terminal domain contains the calcium-independent actin-severin site, whereas the calcium-dependent site is located in the C-terminal portion of the protein. It exhibits significant homology with villin, another calcium-regulated actin filament severing protein. Gelsolin can be found intracellularly, as well as in a secreted form. However, both forms are encoded by the same gene.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Especificaciones

Especificaciones

Gelsolin
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
2
Western Blot
Unconjugated
RUO
Canine, Human, Mouse, Rabbit, Rat
IgG2a
Affinity Purified
Videos
SDS
Documentos

Documentos

Product Certifications

For Research Use Only.