Learn More
Novus Biologicals™ GDI2 Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-11030
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18216135
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
The GDI2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC2596 and based on accession number NM_1494. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens GDP dissociation inhibitor 2 (GDI2), transcript variant 1, mRNA.
Native