missing translation for 'onlineSavingsMsg'
Learn More

GDAP1L1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11977-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16819413

  • 109.00€ / 20 microlitros

Envío estimado: 20-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The ganglioside GD3 synthase causes cell differentiation with neurite sprouting when transfected into the mouse neuroblastoma cell line Neuro2a. After differentiation, the expression of several genes is upregulated, including one that encodes a protein termed ganglioside-induced differentiation-associated protein 1 (Gdap1). A similar gene was found in humans, and mutations in the human gene are associated with Charcot-Marie-Tooth type 4A disease. The protein encoded by this gene is similar in sequence to the human GDAP1 protein. Several transcript variants encoding different isoforms, as well as a noncoding transcript variant, have been found for this gene.
Especificaciones

Especificaciones

GDAP1L1
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VE33, Q96MZ0
GDAP1 L1, GDAP1L1
Rabbit
IgG
20 μL
-20°C
Primary
228858, 78997
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GDAP1L1
Liquid
GDAP1L1
GDAP1L1 Fusion Protein Ag2602
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados