missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GCLM (Human) Recombinant Protein
Human GCLM full-length ORF ( AAH41809, 1 a.a. - 274 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002730-P01.10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Sequence: MGTDSRAAKALLARARTLHLQTGNLLNWGRLRKKCPSTHSEELHDCIQKTLNEWSSQINPDLVREFPDVLECTVSHAVEKINPDEREEMKVSAKLFIVESNSSSSTRSAVDMACSVLGVAQLDSVIIASPPIEDGVNLSLEHLQPYWEELENLVQSKKIVAIGTSDLDKTQLEQLYQWAQVKPNSNQVNLASCCVMPPDLTAFAKQFDIQLLTHNDPKELLSGASFQEALQESIPDIQAHEWVPLWLLRYSVIVKSRGIIKSKGYILQAKRRGS
Especificaciones
AAH41809 | |
glutamate-cysteine ligase, modifier subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GCLM | |
GST |
2730 | |
Wheat germ expression system | |
10 μg | |
GLCLR | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |