missing translation for 'onlineSavingsMsg'
Learn More

GBA2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA554955

Detalles adicionales : Peso : 0.02000kg

Código de producto. 15762881

  • 510.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Immunogen sequence: HDTADWKDLN LKFVLQVYRD YYLTGDQNFL KDMWPVCLAV MESEMKFDKD HDGLIENGGY ADQTYDGWVT TGPSAYCGGL WLAAVAVMVQ MAALCGAQDI QDKFSSILSR GQEAYERLLW NGRYYNYDSS SRPQSRSVMS DQCA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 95%.

GBA2 Polyclonal antibody specifically detects GBA2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Especificaciones

Especificaciones

GBA2
Polyclonal
Unconjugated
GBA2
3.2.1.45; AD035; Beta-glucocerebrosidase 2; beta-glucosidase 2; bile acid; bile acid beta-glucosidase; Bile acid beta-glucosidase GBA2; Bile acid glucosyl transferase GBA2; Cholesterol glucosyltransferase GBA2; Cholesteryl-beta-glucosidase GBA2; F630034E04; GBA2; glucosidase beta 2; glucosidase, beta (bile acid) 2; glucosylceramidase 2; glucosylceramidase beta 2; Kiaa1605; NLGase; non-lysosomal glucosylceramidase; SPG46
Rabbit
Antigen affinity chromatography
RUO
57704
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9HCG7
GBA2
Recombinant protein corresponding to Human GBA2
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Research Use Only