missing translation for 'onlineSavingsMsg'
Learn More

gamma Tubulin Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15176-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814244

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody can label the centrosome structure.

This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
Especificaciones

Especificaciones

gamma Tubulin
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23258, P83887, P83888, Q9GKK5
Gamma 1 tubulin, gamma Tubulin, GCP 1, TUBG, TUBG1, TUBGCP1, Tubulin gamma 1 chain, tubulin, gamma 1
Rabbit
IgG
20 μL
-20°C
Primary
103733, 252921, 403695, 7283
Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
TUBG1
Liquid
TUBG1
gamma Tubulin Fusion Protein Ag7070
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Canine, Human, Mouse, Rat
Videos
SDS
Documentos
Certificados