Learn More
Fisherbrand™ Gamma Radiation Safe Storage Bin
Product Code. 12397107 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Capacity:
1 L
2 L
15 L
20 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
12397107 1 L Each
12317117 20 L Each
12387107 15 L Each
12337117 2 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12397107
Fecha estimada de envoi: 02-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Provides radiation protection shielding for use in laboratories.
  • Serves as solution for short-term storage of radioactive waste or radioisotopes
Specifications
For Use With (Equipment) Bag dimension: L x W x H: 10 x 8 x 13cm (3.93 x 3.14 x 5.11 in.)
Product Type Storage bin
Capacity 1 L