Learn More
Brady™ Galvanized Steel Lockout Cable
Product Code. 11771885 Shop All Brady Products
Change view
Click to view available options
Length (Metric):
2.44 m
3.05 m
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11771885 3.05 m Each
11761885 2.44 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11771885 Supplier Brady™ Supplier No. 065320
  • ONLINE EXCLUSIVE PRICE
    25.40€
    22.16€ / paquete de 1
    Ahorro 3.24€
    12% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Additional Galvanized Steel Lockout Cable.
  • Additional cable.
Specifications
Product Type Lockout Cable
Color Silver
Material Galvanized Steel
Description Dimensions: 3.2 dia. x 3000mmL
Diameter (English) 0.13 in.
Diameter (Metric) 3.18 mm
Length (English) 10.01 ft.
Length (Metric) 3.05 m
Quantity 1 Ea.