Learn More
Emma Safety Footwear Galus S3 Duo Bond Boot
Product Code. 17592466
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17592466 39 1 pair
17572466 37 1 pair
17582466 38 1 pair
17502476 40 1 pair
17512476 41 1 pair
17522476 42 1 pair
17532476 43 1 pair
17542476 44 1 pair
17552476 45 1 pair
17562476 46 1 pair
17572476 47 1 pair
17582476 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 17592466 Supplier Emma Safety Footwear Supplier No. MM58086639
  • ONLINE EXCLUSIVE PRICE
    191.22€
    137.97€ / 1 pares
    Ahorro 53.25€
    27% Descuento
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Insulated with fine quality sheepskin, there is no warmer shoe than the Galus to wear in the coldest of conditions.
  • A naturally better boot.
  • Even in this age of hi-tec, high performance synthetics, sometimes nature's materials are still the best. Which is why we've insulated the Galus with fine quality sheepskin. There's no warmer shoe to wear in the coldest of conditions. And if the temperature suddenly rises up to 300 degrees on the outside, it's reassuring to know the nitrile rubber outsole won't yield one bit.
  • Industries:
    • Chemical industry
    • Construction & technology
  • Safety shoe
    • S3-Safetyshoe with sole plate for wet work
Specifications
Certifications/Compliance ISO 20345:2011
Closure Type Zipper
Color Black
For Use With (Application) Chemical Industry, Construction and Technology
Liner Material Sheepskin
Material Leather
Size 39
Sole Features GripForce™ Duo
Sole Material Nitrile Rubber
Toe Material Steel
Fit Extra-wide Width
EN ISO 20345 S3
Product Type Safety Boot