Learn More
Óxido de galio(III), Puratronic™, 99,999 % (base metálica), Thermo Scientific Chemicals
Descripción
Gallium(III) oxide is used in vacuum deposition. It is useful for making semiconductor devices, gallium-alumina catalyst, gas sensors, luminescent phosphors and dielectric coatings of solar cells. It shows potential for developing deep-ultraviolet TCOs (Transparent Conductive Oxides) and transparent electrodes for ultraviolet optoelectronic devices. Recent studies report that gallium oxide can be a strong contender for power electronic devices for example, in ultrahigh-voltage power switching applications. Films made of gallium oxide have gained commercial interest owing to their gas sensitive characteristics, and glasses made with gallium oxide are the preferred optical materials for use in advanced technologies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Gallium(III) oxide |
Punto de fusión | 1900°C |
Densidad | 6.44 |
Envase | Frasco de plástico |
Índice de refracción | 1.92 |
Cantidad | 5 g |
Gravedad específica | 6.44 |
Información de solubilidad | Soluble in acids. Insoluble in water |
Formula Weight (peso de la fórmula) | 187.44 |
Porcentaje de pureza | 99.999% |
Mostrar más |
RUO – Research Use Only