missing translation for 'onlineSavingsMsg'
Learn More

Galectin 9 Rat anti-Mouse, PerCP-eFluor™ 710, Clone: RG9-35, eBioscience™

Rat Monoclonal Antibody

Marca:  Invitrogen 46-9211-80

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933284

  • 138.00€ / 25 microgramos


Fecha de envío estimada
del proveedor 05-02-2020
Añadir a la cesta

Descripción

Descripción

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin′s disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
Especificaciones

Especificaciones

Galectin 9
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O08573
ecalectin; Gal-9; HUAT; LGALS9A; tumor antigen HOM-HD-21; urate transporter/channel protein, galectin-9
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
RG9-35
PerCP-eFluor™ 710
Lgals9
Liquid
Lgals9
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16859
Videos
Documentos