Learn More
Galectin 9 Rat anti-Mouse, PerCP-eFluor™ 710, Clone: RG9-35, eBioscience™
Rat Monoclonal Antibody
131.00€ - 304.00€
Productos 2
Código de producto Marca Cantidad. Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad. Precio Cantidad y disponibilidad  
15933284 Ver documentos Invitrogen
46-9211-80
25 μg 131.00€
25 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 04-11-2019
Añadir a la cesta
15943284 Ver documentos Invitrogen
46-9211-82
100 μg 304.00€
100 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 04-11-2019
Añadir a la cesta
Descripción
Descripción
Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin′s disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.