missing translation for 'onlineSavingsMsg'
Learn More

Galectin 9 Rat anti-Mouse, APC, Clone: RG9-35, eBioscience™

Rat Monoclonal Antibody

Marca:  Invitrogen 17-9211-80

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923224

  • 96.75€ / 25 microgramos


Fecha estimada de envío
18-08-2020
Añadir a la cesta

Descripción

Descripción

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin′s disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
Especificaciones

Especificaciones

Galectin 9
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O08573
ecalectin; Gal-9; HUAT; LGALS9A; tumor antigen HOM-HD-21; urate transporter/channel protein, galectin-9
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
RG9-35
APC
Lgals9
Liquid
Lgals9
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16859
Videos
Documentos