missing translation for 'onlineSavingsMsg'
Learn More

Galectin-1 Antibody (GAL1/2499R), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79855AF405

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214086

  • 599.34€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Galectin-1 Monoclonal specifically detects Galectin-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Galectin-1
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
LGALS1
Recombinant fragment (around aa 12-108) of human Galectin-1 protein (exact sequence is proprietary) (Uniprot: P09382)
0.1 mL
Apoptosis, Biologically Active Proteins
Human
IgG
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
GAL1/2499R
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1
Rabbit
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales