missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galacton-Star™ Substrate (50X Concentrate)

Concentrated chemiluminescent substrate for E.coli β-galactosidase assays. Thermo Scientific Galacton-Star Substrate (50X Concentrate) is highly sensitive which makes it ideal for detection of weak expression and for transfection normalization with o

Marca:  Invitrogen™ T2264

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 12634345

  • 295.80€ / cada uno


Fecha de envío estimada
del proveedor 04-09-2019
Añadir a la cesta

Descripción

Descripción

Galacton-Star substrate is a chemiluminescent substrate designed for the rapid, and sensitive detection of beta-galactosidase in cell lysates. The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity compared to colorimetric beta-galactosidase assays. Light emission from Galacton-Star reaches maximum in 60-90 minutes and remains constant for at least 1 hour. After incubation at room temperature, the signal is measured in a luminometer. As few as 10 femtograms of beta-galactosidase (20,000 molecules) are detectable.
Especificaciones

Especificaciones

Detection of β-galactosidase
Galacton-Star Substrate
Store at 2° to 8°C
Liquid
2mL
Videos
Documentos

For Research Use Only. Not for use in diagnostic procedures.