missing translation for 'onlineSavingsMsg'
Learn More

GADD45B Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA556268

Detalles adicionales : Peso : 0.02000kg

Código de producto. 15734181

  • 512.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Immunogen sequence: AVEELLVAAQ RQDRLTVGVY ESAKLMNVDP DSVVLCLLAI DEEEEDDIAL QIHFTLIQSF CCDNDINIVR VSGMQRLAQL LGEPAETQGT TEARDLHCLL VTNPHTDAWK SHGLVEVASY CEESRGNNQW VPY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

GADD45B Polyclonal antibody specifically detects GADD45B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Especificaciones

Especificaciones

GADD45B
Polyclonal
Unconjugated
GADD45B
AI323528; DKFZP566B133; Gadd45b; GADD45BETA; growth arrest and DNA damage inducible beta; growth arrest and DNA damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible 45 beta; growth arrest and DNA-damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible, beta; MYD118; myeloid differentiation primary response; myeloid differentiation primary response gene 118; myeloid differentiation primary response protein MyD118; Negative growth regulatory protein MyD118; Unknown (protein for MGC:138134)
Rabbit
Antigen affinity chromatography
RUO
4616
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75293
GADD45B
Recombinant protein corresponding to Human GADD45B
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Research Use Only