Learn More
Gabapentin Related Compound A, TRC
Product Code. 30407038
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30407038 1 g 1g
1 options
This item is not returnable. View return policy
Product Code. 30407038 Supplier Toronto Research Chemicals Supplier No. TRCG1172751G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,3-Pentamethylene-4-butyrolactam
CAS 64744-50-9
Purity Grade Notes HPLC
Molecular Formula C9 H15 N O
Quantity 1 g
Synonym 2-Azaspiro[4.5]decan-3-one, Gabapentin Imp. A (EP), 3,3-Pentamethylene-4-butyrolactam
InChI Formula InChI=1S/C9H15NO/c11-8-6-9(7-10-8)4-2-1-3-5-9/h1-7H2,(H,10,11)
SMILES O=C1CC2(CCCCC2)CN1
IUPAC Name 2-azaspiro[4.5]decan-3-one
Molecular Weight (g/mol) 153.22
Formula Weight 153.1154
Percent Purity >95