Learn More
GAB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA552692
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: CTLVPRRISL SGLDNMRTWK ADVEGQSLRH RDKRLSLNLP CRFSPMYPTA SASIEDSYVP MSPQAGASGL GPHCSPDDYI PMNSGSISSP LPELPANLEP PPVNRDLKPQ RKSRPPPLDL RNLSIIREHA SLTRTRTVPC SRTSF Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 28%.
GAB3 Polyclonal antibody specifically detects GAB3 in Human samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
GAB3 | |
Polyclonal | |
Unconjugated | |
GAB3 | |
5930433H21Rik; DOS/Gab family member 3; GAB3; Gab3 scaffolding protein; GRB2 associated binding protein 3; GRB2-associated binder 3; GRB2-associated binding protein 3; GRB2-associated-binding protein 3; Growth factor receptor bound protein 2-associated protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
139716 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WWW8 | |
GAB3 | |
Recombinant protein corresponding to Human GAB3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only