Learn More
G2E3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551610
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: IQAYPEAFCS ILCHKPESLS AKILSELFTV HTLPDVKALG FWNSYLQAVE DGKSTTTMED ILIFATGCSS IPPAGFKPTP SIECLHVDFP VGNKCNNCLA IPITNTYKEF QENMDFTIRN T Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 81%.
G2E3 Polyclonal antibody specifically detects G2E3 in Human samples. It is validated for Immunohistochemistry.Especificaciones
G2E3 | |
Polyclonal | |
Unconjugated | |
G2E3 | |
6030408C04Rik; 9030416F18; AW046379; D930034K21Rik; G2/M phase-specific E3 ubiquitin-protein ligase; G2/M phase-specific HECT-type E3 ubiquitin transferase; G2/M-phase specific E3 ubiquitin ligase; G2/M-phase specific E3 ubiquitin protein ligase; G2e3; hypothetical protein LOC508575; KIAA1333; mKIAA1333; PHD finger protein 7B; PHF7B; RGD1310263 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55632 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L622 | |
G2E3 | |
Recombinant protein corresponding to Human G2E3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only