Learn More
Ansell Edmont™ Marigold™ G07B Nitrile Gloves
Click to view available options
Tamaño:
Small
Medium
Large
X-Large
Descripción
- 21CFR compliant, approved for all food contact
- Raised diamond grip pattern for safe handling of food products
- Tough and durable in use, with the comfort of latex
- Lightweight construction for greater dexterity and ease in use
- Recommended for use in food processing and preparation and janitorial work
Especificaciones
Certificaciones/conformidad | EN 388 EN 374 PT 2 EN 374 PT 3 21CFR 90/128/EEC |
Color | Blue |
Material | Nitrile |
Palm Thickness (Metric) | 0.22 mm |
Tamaño | Medium |
Modelo | 9834 |