missing translation for 'onlineSavingsMsg'
Learn More

G protein alpha Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 1 publication

Marca:  Bio-Techne NBP1-49874

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214531

  • 480.55€ / 0.10 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

G protein alpha Polyclonal specifically detects G protein alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
Especificaciones

Especificaciones

G protein alpha
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Adenylate cyclase-stimulating G alpha protein, AHO, Alternative gene product encoded by XL-exon, Extra large alphas protein, GNAS complex locus, GNAS1GSA, GNASXL, GPSAC20orf45, GSP, guanine nucleotide binding protein (G protein), alpha stimulating activitypolypeptide 1, guanine nucleotide regulatory protein, guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas, MGC33735, NESP, NESP55, neuroendocrine secretory protein, PHP1A, PHP1B, PHP1C, POH, protein ALEX, SCG6, secretogranin VI, XLalphas
Goat
Affinity Purified
RUO
Primary
This antibody is expected to recognize all three reported isoforms NP_000507.1, NP_001070956.1 and NP_536350.2.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA
0.5 mg/mL
Western Blot 0.1 - 0.5 ug/ml, Peptide ELISA Detection limit 1:32000
NP_000507.1
GNAS
Peptide with sequence C-QAARSNSDGEKATK corresponding to internal region according to NP_001070956.1, NP_001070957.1.
0.1 mg
Signal Transduction
2778
Human, Mouse, Rat
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only