Learn More
Vileda Professional™ CE Sweeper and Pan™
Product Code. 17872476
Change view
Click to view available options
Product Type:
Dustpan Squeegee Sweeper
Open Lobby Dustpan
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17872476 Dustpan Squeegee Sweeper Pack of 5
17862476 Open Lobby Dustpan Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17872476 Supplier Vileda Professional™ Supplier No. 119908
  • ONLINE EXCLUSIVE PRICE
    177.00€
    155.96€ / paquete de 5
    Ahorro 21.04€
    11% Descuento
Fecha estimada de envoi: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The CE Sweeper and Pan™ is ideal for ergonomic and efficient liquid removal. The system is hygienic, lightweight, and flexible. Perfect for dry or damp conditions.
  • Sweep up broken glass in your cleanroom quickly and easily
  • No need to change gloves when an item is dropped on the floor
  • Keeps workers from touching the floor within the controlled environment
  • Can be sprayed or wiped down with common disinfectants
  • Stands up easily for storage or can be hung
  • Adjustable bag holder for different size collection bags
Specifications
Product Type Dustpan Squeegee Sweeper
For Use With (Application) To sweep broken Glass, for the lobby dustpans and smooth floors where dirt could be dry or damp
For Use With (Equipment) Janitorial