Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, lock, 3 in/76 mm
Description
Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, locking rod, 3 in/76 mm, side access, stainless steel- Rack for 76 mm/3 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 148 x 141 x 726 (5.8 x 5.6 x 28.6 in)- Max. box number per rack: 9 (9 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks), CryoCube™ FC660 series (max. 36 racks)- For ULT freezer: Innova C580 (max. 288 boxes), Innova C760 (max. 360 boxes), CryoCube™ FC660 series (max. 324 boxes)- With locking rod for increased sample safety
Specifications
Specifications
| Product Type | Freezer Rack |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.