Learn More
Thermo Scientific™ Frames and Caps for Immuno Standard Modules, Immulon,Frame for 1x12
Product Code. 10140675
Change view
Click to view available options
Array:
8 x 12
Unit Size:
Pack of 10
Product Code. Array Unit Size
10140675 8 x 12 Pack of 10
1 options
This item is not returnable. View return policy
Product Code. 10140675 Supplier Thermo Scientific™ Supplier No. 6604
  • ONLINE EXCLUSIVE PRICE
    74.10€
    65.79€ / paquete de 10
    Ahorro 8.31€
    11% Descuento
Fecha estimada de envoi: 23-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.
Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.
  • Polyethylene, stackable frame is marked for lid orientation
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Description Frame for Immulon modules (1 x 12)
Array 8 x 12
Packaging 10 Pk., 10 Cases
Color Clear
Product Type Frame
No. of Wells 96
No. per Case 10
No. per Pack 10
Plate Compatibility Immulon Modules (1 x 12)