missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19672-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specific to the C-term of FOXL2.
FOXL2 is a winged helix/forkhead transcription factor that is highly conserved in human, goat, mouse, and certain aquatic species. Given its cross-species conservation, it appears to play a critical role evolutionarily, likely in ovarian differentiation in mammals and in ovarian somatic cell differentiation and in further follicle development and/or maintece. Immunohistochemical data shows that FOXL2 is a nuclear protein specifically expressed in eyelids and in fetal and adult ovarian follicular cells. It does not undergo any major posttranslational modifications. FOXL2 is also found to be associated with blepharo-phimosis/ptosis/epicanthus inverses syndrome (BPES). There are two forms of BPES. Type I BPES shows eyelid abnormal-ities that are associated with ovarian failure. In type II BPES, only eyelid defects are found. Studies have shown that FOXL2 is expressed in the mesenchyme of developing mouse eyelids and in adult ovarian follicles.Especificaciones
FOXL2 | |
Polyclonal | |
Unconjugated | |
FOXL2 | |
BPES, BPES1, forkhead box L2, Forkhead box protein L2, FOXL2, PFRK, PINTO, POF3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26927, 367152, 668 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88470, P58012 | |
Foxl2 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |