missing translation for 'onlineSavingsMsg'
Learn More

FOXE3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 55301-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816795

  • 109.00€ / 20 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody is specific to FOXE3.

Forkhead Box Protein E3 (FOXE3, forkhead-related protein FKHL12, forkhead-related transcription factor 8) is a forkhead/winged helix transcription factor, which is expressed in the developing lens from the start of lens placode induction and becomes restricted to the anterior proliferating cells when lens fiber differentiation begins.
Especificaciones

Especificaciones

FOXE3
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13461, Q9QY14
ASMD, FKHL12, forkhead box E3, Forkhead box protein E3, FOXE3, FREAC 8, FREAC8
Rabbit
IgG
20 μL
-20°C
Primary
2301, 30923
Western Blot
Unconjugated
FOXE3
Liquid
FOXE3
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
Videos
SDS
Documentos
Certificados