Learn More
Formal Buffered Saline, Reagecon
BIOPHARMA
Product Code. 11593192 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
HDPE Bottle GL45
HDPE Jerrycan
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11593192 5 L HDPE Jerrycan 5 L
11503202 1 L HDPE Bottle GL45 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11593192 Supplier Reagecon Supplier No. FBS5
  • ONLINE EXCLUSIVE PRICE
    54.65€
    39.89€ / 5 litros
    Ahorro 14.76€
    27% Descuento
 Restricted Item
Fecha estimada de envoi: 13-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Fixative mixture of formaldehyde in isotonic saline for routine histopathology.
  • Fixative for histopathology.
Specifications
Physical Form Liquid
Packaging HDPE Jerrycan
Quantity 5 L
compliance-icons compliance-icons
Product Identifier
  • FORMAL BUFFERED SALINE
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contain: Formaldehyde, Sodiumdihydrogenphosphate