Learn More
Gilac™ Food Specimen Containers with Lids
Product Code. 17918531
Change view
Click to view available options
Capacity (Metric):
3 L
8 L
Quantity:
5 Pk.
7 Pk.
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Quantity Unit Size
17918531 8 L 7 Pk. Each
17958671 3 L 5 Pk. Each
17958821 3 L 7 Pk. Each
17948851 8 L 5 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17918531 Supplier Gilac™ Supplier No. G160480
  • ONLINE EXCLUSIVE PRICE
    211.35€
    139.13€ / paquete de 1
    Ahorro 72.22€
    34% Descuento
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
Food Specimen Containers with Lids are designed especially for storing food specimen samples complying with HACCP catering standards. Markings do not wash off, even in the machine.
  • Pack of food specimen containers.
  • Containers especially for storing food specimen samples complying with HACCP catering standards.
  • Markings do not wash off, even in the machine.
Specifications
Product Type HACCP Food Container
Material HDPE
Capacity (English) 2.1 gal.
Capacity (Metric) 8 L
Color Assorted
Height (English) 6.3 in.
Height (Metric) 160 mm
Length (English) 19.3 in.
Length (Metric) 491 mm
Quantity 7 Pk.
Width (English) 13.3 in.
Width (Metric) 337 mm