Learn More
Invitrogen™ Follistatin (FST) Human ELISA Kit
Sandwich ELISA Kit
656.00€ - 4715.00€
Especificaciones
Número de acceso | P19883 |
---|---|
Sensibilidad del ensayo | 500 pg |
ensayo | ELISA sandwich de fase sólida |
Conjugado | Biotina |
Tipo de producto | Kit de ELISA |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15415003
|
Invitrogen™
EHFST |
96 pruebas |
656.00€
1 set |
Fecha estimada de envío: 14-05-2024
Inicie sesión para ver el stock |
|||||
17874200
|
Invitrogen™
EHFSTX10 |
10 × 96 pruebas |
4715.00€
paquete de 10 |
Fecha estimada de envío: 22-05-2024
Inicie sesión para ver el stock |
|||||
17145449
|
Invitrogen™
EHFSTX5 |
5 × 96 pruebas |
2635.00€
cada uno |
Fecha estimada de envío: 21-05-2024
Inicie sesión para ver el stock |
|||||
Descripción
The Human Follistatin (Hu FST) ELISA quantitates Hu FST in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FST. Principle of the method The Human FST solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Follistatin is a single-chain gonadal protein that specifically inhibits follicle-stimulating hormone release. The single FST gene encodes two isoforms, FST317 and FST344 containing 317 and 344 amino acids respectively, resulting from alternative splicing of the precursor mRNA. In a study in which 37 candidate genes were tested for linkage and association with polycystic ovary syndrome (PCOS) or hyperandrogenemia in 150 families, evidence was found for linkage between PCOS and follistatin.Especificaciones
P19883 | |
ELISA sandwich de fase sólida | |
Kit de ELISA | |
Plasma, Sobrenadante, Suero | |
Humano | |
Lector de microplacas colorimétrico | |
FS | |
< 10 % | |
RUO | |
De 2 °C a 8 °C | |
Humano | |
4 h 45 min |
500 pg | |
Biotina | |
Otras proteínas | |
Este kit de ELISA no muestra reactividad cruzada con las siguientes citoquinas probadas: angiogenina humana, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alfa, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF- 7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, leptina (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alfa, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alfa, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alfa, TNF-beta, TPO, VEGF. | |
ELISA, ensayos de proteínas, biología de proteínas, biología del cáncer, inmunología, neurociencia, neurología, detección de proteínas, análisis de proteínas | |
10468 | |
< 12 % | |
Peroxidasa de rábano picante (HRP) | |
Diferenciación | |
FS | |
1 h 20 min |