Learn More
Velp Scientifica™ FOC 120E Cooled Incubator
Velp Scientifica™ FOC 120E Cooled Incubator is a high-efficiency refrigerated thermostat that offers excellent stability and homogeneity of the internal temperature.
Marca:  Velp Scientifica™ F10300310
Detalles adicionales : Peso : 36.00000kg
Código de producto. 15309474
  • 1750.00€ / cada uno
Fecha de envío estimada
del proveedor 17-05-2019
Añadir a la cesta
Descripción
Descripción
  • Provided with the innovative Auto-Tuning thermoregulation system, and forced air circulation
  • Offer excellent stability and homogeneity of the internal temperature
Especificación
Especificación
3 to 50°C
912 mm
50/60 Hz
768 mm
No
230 V, 50/60 Hz
2
36 kg
±0.5°C
120L
230 V
550 mm
392 mm
550 x 540 x 912 mm
3-digit Digital
540 mm
FOC 120E
Videos