Learn More
Thermo Scientific™ Fluid Transfer Assembly, Pump Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Marca:  Thermo Scientific™ SH31113.01
Detalles adicionales : Peso : 0.70000kg
 Ver más versiones de este producto
Código de producto. 15929866
  • 38.63€ / 1 unidad
Fecha de envío estimada
del proveedor 16-03-2020
Añadir a la cesta
Descripción
Descripción
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Especificaciones
Especificaciones
Pump Set
12.7mm (1/2 in.) ID x 19.05mm (3/4 in.) OD; 0.25m(10 in.) C-Flex 374 to 0.5 m (19.7 in.) Pharmed BPT to 0.25m(10 in.); C-Flex 374; MPX