Learn More
Abnova™ FLT1 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human FLT1
Marca:  Abnova™ KA0665.1Kit
Detalles adicionales : Peso : 0.02000kg
 Ver más versiones de este producto
Código de producto. 16056205
  • 578.00€ / 1 set
Fecha estimada de envío
20-02-2020
Añadir a la cesta
Especificaciones
Especificaciones
31.2 to 2000pg/mL
Human
Colorimetric
96 wells Antibody coated microwell, 10ng lyophilized recombinant Mouse Fn1 standard vials, 30mL sample diluent Buffer, 130μL Biotinylated Anti-Mouse Fn1 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin Biotin Peroxidase c
FLT1 (Human) ELISA kit
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
96 Assays
Biotin
sFLT1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human soluble FLT1.
Quantification
sFLT1 (Human) ELISA Kit
1 Kit
Store the kit at-20°C.
100μL