missing translation for 'onlineSavingsMsg'
Learn More

Flt-3/Flk-2/CD135 Antibody (12B6), FITC, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Bio-Techne NB110-94155F

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213501

  • 590.70€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Flt-3/Flk-2/CD135
Monoclonal
FITC
PBS with 0.05% Sodium Azide
FLT3
Mouse Flt-3/Flk-2/CD135 (extracellular domain)
0.1 mL
Primary
This recognizes mouse Flt3/CD135 Antibody (12B6) in western blot.
Store at 4C in the dark.
IgG2
Immunohistochemistry, Immunohistochemistry (Paraffin)
12B6
Immunohistochemistry, Immunohistochemistry-Paraffin
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Rat
Protein G purified
Cell Cycle and Replication, Phospho Specific
2322
Mouse
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales